EMAIL THIS PAGE TO A FRIEND
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova
[Comparative study of himantane and amantadine effects in experimental intracerebral posttraumatic hematoma].
PMID 23235415
Abstract
Effects of the novel antiparkinsonian drug himantane and amantadin were studied in rats with intracerebral posttraumatic hematoma. Drugs were administered first at 3.5 hours after surgery and then for 4 consecutive days. Effects were registered on days 1, 3, 7 and 14 after surgery. It was shown that both drugs significantly decreased mortality and improved motor activity, exploratory behavior and memory. Amantadin was more effective in tests for motor activity and exploratory behavior. Himantane 5 mg/kg i.p demonstrated the more pronounced activity in restoring memory. The results obtained testify for neuroprotective properties of the novel antiparkinsonian drug himantane.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1260
Amantadine hydrochloride
C10H17N · HCl