EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Preparation and study of an f,f,f',f'' covalently linked tetranuclear hetero-trimetallic complex - a europium, terbium, dysprosium triad.
PMID 23236602
Abstract
A heterotrimetallic tetranuclear lanthanide complex containing two dysprosium ions, a terbium ion and a europium ion has been prepared by coupling three kinetically stable complexes together using the Ugi reaction. The covalently linked trimetallic system exhibits luminescence from all the different lanthanide centres.