EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Study on chemical constituents from Anoectochilus chapaensis].
PMID 23236826
Abstract
To investigate the chemical constituents from the whole herbs of Anoectochilus chapaensis. The chemical constituents were isolated and purified by repeated column chromatographies with silica gel, Sephadex LH-20 and preparative TLC. Their structures were identified by their physiochemical property and spectral data. 6 compounds were isolated and elucidated as follows: (1) Friedelin, (2) Sorghumol, (3) 5alpha, 8alpha-epidioxyergost-22-en-3beta-ol, (4) Stearic acid, (5) Octadecane and (6) Epifriedelanol. Compound (1), (2) and (4) are isolated from this plant for the first time, and compound (3), (5) and (6) are isolated from genera Anoectochilus for the first time.