EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Chemical constituents of the roots of Vaccinium bracteatum].
PMID 23236828
Abstract
To study the chemical constituents of the roots of Vaccinium bracteatum. The constituents were separated and purified with chromatographic methods (including silica gel, Sephadex LH-20 and RP-18 column chromatography), and their structures were determined by spectroscopic methods (including MS, 1H-NMR and 13C-NMR). 10 compounds were isolated from the roots of Vaccinium bracteatu and were elucidated as chlorogenic acid (1), pinoresinol (2), ferulic acid (3), kaempferol (4), trans-caffeic acid (5), beta-sitosterol (6), quercetin (7), oleanolic acid (8), apigenin (9) and luteolin (10). Compounds 1 -3 are obtained from this plant for the first time.