EMAIL THIS PAGE TO A FRIEND
Chemosphere
The impact of chlorinated solvent co-contaminants on the biodegradation kinetics of 1,4-dioxane.
PMID 23237300
Abstract
1,4-Dioxane (dioxane), a probable human carcinogen, is used as a solvent stabilizer for 1,1,1-trichloroethane (TCA) and other chlorinated solvents. Consequently, TCA and its abiotic breakdown product 1,1-dichloroethene (DCE) are common co-contaminants of dioxane in groundwater. The aerobic degradation of dioxane by microorganisms has been demonstrated in laboratory studies, but the potential effects of environmentally relevant chlorinated solvent co-contaminants on dioxane biodegradation have not yet been investigated. This work evaluated the effects of TCA and DCE on the transformation of dioxane by dioxane-metabolizing strain Pseudonocardia dioxanivorans CB1190, dioxane co-metabolizing strain Pseudonomas mendocina KR1, as well as Escherichia coli expressing the toluene monooxygenase of strain KR1. In all experiments, both TCA and DCE inhibited the degradation of dioxane at the tested concentrations. The inhibition was not competitive and was reversible for strain CB1190, which did not transform the chlorinated solvents. For both strain KR1 and toluene monooxygenase-expressing E. coli, inhibition of dioxane degradation by chlorinated solvents was competitive and irreversible, and the chlorinated solvents were degraded concurrently with dioxane. These data suggest that the strategies for biostimulation or bioaugmentation of dioxane will need to consider the presence of chlorinated solvents during site remediation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48614 1,1,1-Trichloroethane solution, certified reference material, 200 μg/mL in methanol
40010-U 1,1,1-Trichloroethane solution, certified reference material, 5000 μg/mL in methanol
40027 1,1-Dichloroethene solution, certified reference material, 1000 μg/mL in methanol
C2H2Cl2
163023
1,1-Dichloroethylene, contains 200 ppm MEHQ as inhibitor, 99%
C2H2Cl2
02574
1,1-Dichloroethylene, analytical standard
C2H2Cl2
360481
1,4-Dioxane, ACS reagent, ≥99.0%
C4H8O2
34857
1,4-Dioxane, for HPLC, ≥99.5%
C4H8O2
D201863
1,4-Dioxane, ReagentPlus®, ≥99%
C4H8O2
33147
1,4-Dioxane, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
C4H8O2
676934
1,4-Dioxane, ACS reagent, ≥99.0%
C4H8O2
CRM48367
1,4-Dioxane solution, certified reference material, 2000 μg/mL in methanol, ampule of 1 mL
C4H8O2
551368
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D)
C4H8O2
736171
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 5 mm × 8 in.
C4H8O2
704806
1,4-Dioxane solution, NMR reference standard, 1 mM in D2O ("100", 99.96 atom % D), NMR tube size 5 mm × 7 in.
C4H8O2
704814
1,4-Dioxane solution, NMR reference standard, 10 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.
C4H8O2
704822
1,4-Dioxane solution, NMR reference standard, 0.1 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.
C4H8O2
703354
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 6.5 mm × 8 in.
C4H8O2
684228
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 10 mm × 8 in.
C4H8O2
631868
1,4-Dioxane solution, NMR reference standard, 5 mM in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
C4H8O2
631833
1,4-Dioxane solution, NMR reference standard, 5 mM in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
C4H8O2
611905
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm × 8 in.
C4H8O2
612073
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 3 mm × 8 in.
C4H8O2
612111
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
C4H8O2