EMAIL THIS PAGE TO A FRIEND
Malaria journal
Rationale for recommending a lower dose of primaquine as a Plasmodium falciparum gametocytocide in populations where G6PD deficiency is common.
PMID 23237606
Abstract
In areas of low malaria transmission, it is currently recommended that a single dose of primaquine (0.75 mg base/kg; 45 mg adult dose) be added to artemisinin combination treatment (ACT) in acute falciparum malaria to block malaria transmission. Review of studies of transmission-blocking activity based on the infectivity of patients or volunteers to anopheline mosquitoes, and of haemolytic toxicity in glucose 6-dehydrogenase (G6PD) deficient subjects, suggests that a lower primaquine dose (0.25 mg base/kg) would be safer and equally effective. This lower dose could be deployed together with ACTs without G6PD testing wherever use of a specific gametocytocide is indicated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G6378 Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
G7877 Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
G4134 Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
G5885 Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, 550-1,100 units/mg protein (biuret)
G8404 Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret)
G5760 Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret
G8529 Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, lyophilized powder, 550-1,100 units/mg protein (biuret)
G2921
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, BioUltra, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥95% (SDS-PAGE), ≥550 units/mg protein (biuret)
G8289 Glucose-6-phosphate dehydrogenase from torula yeast, ammonium sulfate suspension, 300-600 units/mg protein (biuret)
160393
Primaquine bisphosphate, 98%
C15H21N3O · 2H3PO4