EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
L-amino acid biosensor based on L-amino acid oxidase immobilized onto NiHCNFe/c-MWCNT/PPy/GC electrode.
PMID 23237796
Abstract
A method is described for construction of a highly sensitive amperometric L-amino acid biosensor based on covalent immobilization of goat kidney L-amino acid oxidase (LAAO) onto carboxylated multiwalled carbon nanotubes/nickel hexacyanoferrate/polypyrrole hybrid film electrodeposited on surface of glassy carbon electrode. The biosensor showed optimum response within 5s at pH 7.5 and 35 °C, when polarized at 0.15 V vs. Ag/AgCl. There was a linear relationship between biosensor response (μA) and L-phenylalanine concentration in the range, 0.5 μM to 100 mM. The sensitivity of the biosensor was 79.31 nA cm(-2) μM(-1) with a detection limit of 0.5 μM (S/N=3). The enzyme electrode was used 160 times over a period of 140 days, when stored dry at 4 °C. The biosensor was evaluated and employed for determination of L-amino acid level in fruit juices and alcoholic beverages. The biosensor showed excellent analytical characteristics along with short response time, good shelf life, and negligible interference of commonly present interferents. The film could be used for improvement of other biosensors also.