EMAIL THIS PAGE TO A FRIEND
Nanotechnology
V-groove SnO2 nanowire sensors: fabrication and Pt-nanoparticle decoration.
PMID 23237979
Abstract
Networked SnO(2) nanowire sensors were achieved using the selective growth of SnO(2) nanowires and their tangling ability, particularly on on-chip V-groove structures, in an effort to overcome the disadvantages imposed on the conventional trench-structured SnO(2) nanowire sensors. The sensing performance of the V-groove-structured SnO(2) nanowire sensors was highly dependent on the geometrical dimension of the groove, being superior to those of their conventional trench-structured counterparts. Pt nanoparticles were decorated on the surface of the networked SnO(2) nanowires via γ-ray radiolysis to enhance the sensing performances of the V-groove sensors whose V-groove widths had been optimized. The V-groove-structured Pt-nanoparticle-decorated SnO(2) nanowire sensors exhibited outstanding and reliable sensing capabilities towards toluene and nitrogen dioxide gases, indicating their potential for use as a platform for chemical gas sensors.