EMAIL THIS PAGE TO A FRIEND
Pharmacological reports : PR
Synergistic neurotoxicity of oxygen-glucose deprivation and tetrabromobisphenol A in vitro: role of oxidative stress.
PMID 23238473
Abstract
Tetrabromobisphenol A (TBBPA) is a toxic brominated flame retardant. Previous studies have demonstrated that exposure of primary cultures of rat cerebellar granule cells (CGC) to ≥ 10 μM TBBPA induces toxicity and excitotoxicity, and the underlying mechanism may involve calcium imbalance and oxidative stress. Here we examined whether the application of TBBPA at subtoxic concentrations may exacerbate acute damage of CGC challenged with oxygen-glucose deprivation (OGD), and evaluated with fluorescent indicators the involvement of calcium imbalance, mitochondrial depolarization and oxidative stress. Survival of CGC was assessed 24 h after OGD/TBBPA using fluorescent dyes. An OGD challenge lasting for 45, 60 or 75 min induced a duration-dependent injury to the neurons. Application of 2.5, 5 or 7.5 μM TBBPA for 45 min to normoxic and glucose-containing incubation medium did not reduce the viability of cultured CGC, but this compound exacerbated the toxic effects of OGD in a concentration-dependent way. Moreover, TBBPA had a slight effect on calcium homeostasis and mitochondrial membrane potential, but significantly activated the production of reactive oxygen species in CGC. The application of H(2)O(2) at 5, 10 and 25 μM mimicked the effects of TBBPA on OGD toxicity, while 0.1 mM ascorbic acid or 1 mM glutathione ameliorated this toxicity. These results suggest the involvement of oxidative stress in the synergistic neurotoxic effects of TBBPA and OGD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11223
3,3′,5,5′-Tetrabromobisphenol A, analytical standard
C15H12Br4O2