EMAIL THIS PAGE TO A FRIEND
The Tokai journal of experimental and clinical medicine
Pharmacological conversion of atrial fibrillation in the patients of Graves' disease.
PMID 23238902
Abstract
Hyperthyroidism is one of the common causes of atrial fibrillation (AF), and AF is associated with increased morbidity and mortality due to thromboembolism. The sinus rhythm maintenance rate of hyperthyroidism-induced AF patients after conversion to sinus rhythm is excellent. The present study was undertaken to assess the efficacy and safety of bepridil, a multichannel blocker, in patients with hyperthyroidism-induced persistent AF. Sixty-two patients with hyperthyroidism-induced persistent AF were treated with bepridil. Oral bepridil therapy resulted in conversion to sinus rhythm in 32 (51.6%) of the 62 patients. There were no significant differences in clinical characteristics between the responders and non-responders. At the observation period of an average of 23.9 months, the sinus rhythm maintenance rate was found to be 81.3%. Adverse effects consisted of abnormal QTc prolongation in 3 patients and sinus bradycardia in 10 patients. There was one death in which a causal association with bepridil could not be ruled out. Bepridil is as beneficial treatment to convert AF for the patients with hyperthyroidism-induced persistent AF as it is for the patients with AF due to other causes. However, bepridil should be used with caution to avoid serious side effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5016
Bepridil hydrochloride, powder
C24H34N2O · HCl