EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
A novel approach to the analysis of SUMOylation with the independent use of trypsin and elastase digestion followed by database searching utilising consecutive residue addition to lysine.
PMID 23239325
Abstract
Identification of sites of protein SUMOylation is of great importance due its functional diversity within the cell. To date, most approaches to this problem rely on site-directed mutagenesis and/or highly specialised mass spectrometry approaches. We present a novel alternative approach to the site mapping of SUMOylation using trypsin and elastase digestion, routine mass spectrometry and an unbiased isotag database searching strategy. SUMOylated protein samples were digested with a number of enzymes and the resulting peptides separated using liquid chromatography. Analysis was carried out on both linear ion trap Orbitrap and quadrupole-time-of-flight (Q-TOF)-based mass spectrometers equipped with electrospray ionisation. The data files were subsequently searched using the Mascot algorithm with multiple variable tag modifications corresponding to SUMO-derived fragments. The utility of this approach was demonstrated with di-SUMO 2, di-SUMO 3, SUMO 1-RanGap(418-587) 1 and an enriched population of SUMOylated proteins. Unbiased database searches led to the identification of a number of analytically useful isotags ranging in length from two to four residues. Isopeptide fragments were generated including QTGG (di-SUMO-2/3), TGG (di-SUMO-2/3) and GG (SUMO-1). The method was validated by successfully mapping a number of sites of SUMO modification on SUMO-modified proteins enriched from a cell lysate. This combination of relaxed enzyme specificity, shortened isotag generation and unbiased database searching enabled confident identification of novel analytically useful SUMOylated isopeptides without a requirement for mutagenesis.