EMAIL THIS PAGE TO A FRIEND
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences
[Anticancer effect of triptolide-polyethylenimine-cyclodextrin in vitro].
PMID 23239652
Abstract
To develop a drug delivery system triptolide-polyethylenimine-cyclodextrin and to evaluate its anticancer activity in vitro. Triptolide was conjugated to polyethylenimine-cyclodextrin by N, N'-carbonyldiimidazole to form triptolide-polyethylenimine-cyclodextrin. (1)H-NMR, FT-IR and XRD were used to confirm its structure. The anticancer effect of the polymer was assessed by MTT assay, erasion trace test and hematoxylin-eosin staining. The potential to condense siRNA and to delivery siRNA into cytoplasm was demonstrated by gel retardation assay, zeta-potential determination and fluorescence staining. Triptolide was successfully conjugated to polyethylenimine-cyclodextrin and the conjugation rate of triptolide was 10% (w/w). siRNA was effectively condensed by the polymer at the N/P ratio of 5, and its particle size was 300 ±15 nm and zeta potential was 8 ±2.5 mV. MTT assay, erasion trace test and hematoxylin-eosin staining revealed that triptolide-polyethylenimine-cyclodextrin had anticancer effect and low cytotoxicity to normal cells. The polymer was able to deliver siRNA to the cytoplasm effectively as demonstrated by fluorescence staining. Triptolide-polyethylenimine-cyclodextrin is able to inhibit the growth and migration of cancer cells in vitro and to carry siRNA into cells effectively. It is potential to be used as a novel prodrug for co-delivery of gene and drug in cancer treatment.