EMAIL THIS PAGE TO A FRIEND
Guang pu xue yu guang pu fen xi = Guang pu
Determination of sulfadiazine based on its derivatization with fluorescamine by self-ordered ring fluorescence microscopic imaging technique.
PMID 23240417
Abstract
A self-ordered ring (SOR) fluorescence microscopic imaging technique has been developed for the determination of trace amounts of sulfadiazine based on its derivatization with fluorescamine. In the presence of HAc-NaAc buffer solution (pH 3.12) and polyvinyl alcohol-124 (PVA-124), the droplet containing fluorescamine derivatized sulfadiazine can form a SOR on the solid support after solvent evaporation with the diameter of 1.86 mm and its ring belt width of 54.9 microm. The quantitative analysis of sulfadiazine is achieved with the linear range of 7.8 x 10(-14)-1.8 x 10(-12) mol x ring(-1) (3.9 x 10(-7)-9.0 x 10(-6) mol x L(-1) and detection limit of 7.8 x 10(-5) mol x ring(-1) (3.9 x 10(-8) mol x L(-1)) when 0.2 microL droplet was spotted. The technique has been satisfactorily applied to the determination of sulfadiazine in the tablet, synthetic sample and residues in six different milk samples with the recoveries of 91.0%-105.8%, respectively, and RSDs less than 4.4%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47614
Fluram, BioReagent, suitable for fluorescence, ≥99.0% (UV)
C17H10O4