EMAIL THIS PAGE TO A FRIEND
Anesthesia progress
Basic and clinical pharmacology of autonomic drugs.
PMID 23241039
Abstract
Autonomic drugs are used clinically to either imitate or inhibit the normal functions of the sympathetic and parasympathetic nervous systems. A large number of additional drug classes also interact with these systems to produce a stunning number of possible side effects. This article reviews the basic function of the autonomic nervous system and the various drug classes that act within these neural synapses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
134910
(1R,2S)-(−)-Ephedrine, 98%
C10H15NO