EMAIL THIS PAGE TO A FRIEND
Rheumatology (Oxford, England)
Treatment of fracture non-union in a young adult with combination anabolic and anti-resorptive bone therapy.
PMID 23241394
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3796 Parathyroid Hormone Fragment 1-34 human, ≥95% (HPLC), powder
C181H291N55O51S2