EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
Case report: Transient success using prolonged treatment with miltefosine for a patient with diffuse cutaneous leishmaniasis infected with Leishmania mexicana mexicana.
PMID 23243111
Abstract
Leishmania sp. is an intracellular parasite that causes a variable degree of clinical manifestations, especially in the skin. We present the case of a 38-year-old male with a chronic history of mucocutaneous disease present since childhood that generated deformity, loss of cartilage in the ears and nose, and scarring that limited his range of motion. The parasite was identified as L. mexicana mexicana. The patient was treated with a 3-month course of oral miltefosine with overwhelming results.