EMAIL THIS PAGE TO A FRIEND
Przeglad lekarski
[Cardiac arrest during intoxication with nicotine--a case report and a review of literature].
PMID 23243942
Abstract
There are numerous cases of acute intoxication with nicotine described in medical literature. The known routes of intoxication are transdermal, oral, inhalational, and intravenous. We presents a case of oral poisoning with nicotine from cartridges of e-cigaretes. According to the best of our knowledge this is first case of such poisoning in Poland. The intoxication had serious course, complicated with sudden cardiac arrest.