EMAIL THIS PAGE TO A FRIEND
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology
Effect of oral administration with pravastatin and atorvastatin on airway hyperresponsiveness and allergic reactions in asthmatic mice.
PMID 23244652
Abstract
Asthma is characterized by airway hyperresponsiveness and remodeling. Pravastatin and atorvastatin are used clinically as cholesterol-lowering agents but also exhibit anti-inflammatory and immunomodulating properties. To investigate the therapeutic effect of oral statins on airway hyperresponsiveness and allergic reaction. BALB/c mice received intraperitoneal sensitization and aerosol inhalation with ovalbumin consequently. One week after ovalbumin aerosol challenge, pravastatin, atorvastatin, or phosphate-buffered saline were given by intragastric gavage daily for 2 weeks. Airway hyperresponsiveness, serum allergen specific antibody levels, cytokine production by splenocytes, and bronchoalveolar lavage fluid were examined. Both pravastatin and atorvastatin effectively reduced airway hyperresponsiveness. Pravastatin effectively suppressed both T(H)1- and T(H)2-mediated antibody responses, reducing serum specific IgE, IgG, IgG1, and IgG2a levels. Pravastatin also effectively reduced interleukin (IL) 4, IL-5, and interferon γ production but significantly enhanced IL-10 levels in splenocytes and BALF. Similarly, atorvastatin effectively attenuated production of specific IgE, IgG1, and IgG2a antibodies. It also significantly attenuated IL-4, interferon γ, and increased IL-10 concentration in bronchoalveolar lavage fluid and splenocytes. Oral administration of pravastatin or atorvastatin not only was able to inhibit T(H)1 inflammatory responses but also had therapeutic effects on airway hyperresponsiveness and T(H)2 allergic responses. These results seem to suggest that these drugs have potential as a nonimmunosuppressive therapy for asthma and allergic diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4498
Pravastatin sodium salt hydrate, ≥98% (HPLC), powder
C23H35O7Na · xH2O