EMAIL THIS PAGE TO A FRIEND
Journal of photochemistry and photobiology. B, Biology
Acute stress in seedlings detected by ultra-weak photon emission.
PMID 23245730
Abstract
The ultra-weak photon emission (biophoton) patterns of wheat germination tests under chemical stress (mercury and fluoride solutions) are presented in comparison to respective control groups run simultaneously in two similar, automatic photon-count chambers. Solutions of Hg (EC(50) and EC(100)) and NaF (EC(50)) were applied at the fourth day, after the first 72 h of imbibition in water, with photon-count experiments in controlled conditions. Seedlings' growth performance were checked by total mass increase as well as root development, and plotted against the respective angular factor of photon-count curve's linear regression. Both Hg and NaF series presented very distinguishable patterns when compared to each respective control groups. Higher counts appear all over the 24 h test for the Hg series and just in the first 6 h for the NaF series.