EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Cyanide modulated fluorescent supramolecular assembly of a hexaphenylbenzene derivative for detection of trinitrotoluene at the attogram level.
PMID 23247388
Abstract
Fluorescent nanorods formed by self-assembly of hexaphenylbenzene derivative in the presence of cyanide ions serve as a sensitive colorimetric and fluorogenic sensor for the detection of trinitrotoluene (TNT) at the attogram (10(-18) g) level with a detection limit of 10.21 ppq (parts per quadrillion).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
149454
Hexaphenylbenzene, 98%
C42H30