EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Structure of amiprilose hydrochloride, a novel anti-inflammatory agent.
PMID 2324965
Abstract
Amiprilose hydrochloride is a carbohydrate-derived, novel anti-inflammatory with potential application in the treatment of rheumatoid arthritis. A spectroscopy-based approach was undertaken to assign both the relative and absolute configuration of its five chiral centers. The fully assigned 13C and 1H NMR spectra of amiprilose hydrochloride was used to establish the relative stereochemistry of four of its five chiral centers held rigid in its furanose ring system. Parallel synthesis of the enantiomer of amiprilose hydrochloride from L-glucose was followed by CD spectropolarimetry to establish that no inversion of chiral centers had occurred in the synthesis. The hydrobromide salt of amiprilose and its enantiomer were prepared and, together with amiprilose hydrochloride, were crystallized. X-ray crystallographic analysis resulted in the assignment of the absolute configuration of all five chiral centers.