EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
Cryptic and asymptomatic Opisthorchis felineus infections.
PMID 23249682
Abstract
We describe the diagnostic difficulties experienced during an opisthorchiasis outbreak. Of 31 infected individuals, 61.3% were asymptomatic, and in the 12 symptomatic individuals, the duration of non-pathognomonic symptoms was shorter than 4 weeks. Serology by enzyme-linked immunosorbent assay and polymerase chain reaction fecal analysis were shown to be the most sensitive diagnostic tools.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2523
Mebendazole, analytical standard, ≥98% (HPLC)
C16H13N3O3