EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[GC-MS analysis of chemical components in seeds oil from Croton tiglium].
PMID 23252276
Abstract
To identify the chemical components and their relative content in seeds oil from Croton tiglium. The oil obtained by extracting of the seeds of Croton tiglium with petroleum ether was subjected to methyl-esterification or dilution with ethylether. GC-MS were used to identify the components in croton oil,peak area normalization method was used to determine the relative content of these substances in the sample. Seventeen fat acid components were identified from croton oil. The main components were linoleic acid, oleic acid, and eicosenoic acid in methyl-esterified sample, whose quantities accouted for 77.33%. In addition, five aromatic compounds were also found in the sample diluted with ethylether, such as isoborneol, fenchyl alcohol, etc. Phorbol esters, having carcinogenesis and anti-HIV-1 effects, were not been identified. There are abundant of linoleic acid, oleic acid and eicosenoic acid in the seeds oil extracted from Chinese Croton tiglium. In contrast, the active component with carcinogenesis and anti-HIV-1 might be very rare in the samples and difficult to be obtained by ordinary separating and extracting methods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6719 Croton oil