EMAIL THIS PAGE TO A FRIEND
British journal of haematology
Erythrocyte adenosine deaminase: diagnostic value for Diamond-Blackfan anaemia.
PMID 23252420
Abstract
Diamond-Blackfan anaemia (DBA) is an inherited bone marrow failure syndrome (IBMFS) characterized by red cell aplasia. Mutations in ribosomal genes are found in more than 50% of cases. Elevated erythrocyte adenosine deaminase (eADA) was first noted in DBA in 1983. In this study we determined the value of eADA for the diagnosis of DBA compared with other IBMFS; the association of eADA in DBA with age, gender or other haematological parameters; and the association with known DBA-related gene mutations. For the diagnosis of DBA compared with non-DBA patients with other bone marrow failure syndromes, eADA had a sensitivity of 84%, specificity 95%, and positive and negative predictive values of 91%. In patients with DBA there was no association between eADA and gender, age, or other haematological parameters. Erythrocyte ADA segregated with, as well as independent of, known DBA gene mutations. While eADA was an excellent confirmatory test for DBA, 16% of patients with classical clinical DBA had a normal eADA.