EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Diastereoselective one-pot Knoevenagel condensation/Corey-Chaykovsky cyclopropanation.
PMID 23252964
Abstract
Efforts to substitute the cyclopropane ring in a series of aryl cyclopropylnitriles led to the discovery of an operationally simple one-pot method for Knoevenagel condensation and subsequent Corey-Chaykovsky cyclopropanation giving diastereomerically pure products as a racemic mixture of enantiomers. Method development and results for variably substituted aryl acetonitriles and aldehydes in the reaction are reported. A concise synthesis of (±)-bicifadine in two steps is provided to demonstrate the utility of the method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295183
Cyclopropane, ≥99%
C3H6