EMAIL THIS PAGE TO A FRIEND
Organic letters
Diastereoselective intramolecular carbamoylketene/alkene [2 + 2] cycloaddition: enantioselective access to pyrrolidinoindoline alkaloids.
PMID 23252989
Abstract
A novel and highly diastereoselective intramolecular carbamoylketene/alkene [2 + 2] cycloaddition has been developed, and the methodology was successfully applied to the enantioselective syntheses of (-)-esermethole and Takayama's intermediate for (+)-psychotrimine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45710
Eserine, ≥98.0% (N)
C15H21N3O2