EMAIL THIS PAGE TO A FRIEND
Internal medicine journal
Leukaemia cutis in chronic lymphocytic leukaemia following varicella zoster virus reactivation.
PMID 23253001
Abstract
We report a case of a 75-year-old male with indolent chronic lymphocytic leukaemia (CLL) for 8 years, who presented with a 6-month history of a painful, zosteriform eruption in a T3-4 distribution that evolved into an unusual crop of papular nodules. Upon biopsy and immunostaining of these lesions CLL was proven consistent with leukaemia cutis related to varicella-zoster virus reactivation. In the absence of other treatment indices, he was commenced on chlorambucil with successful resolution of both his pain and the lesions.