EMAIL THIS PAGE TO A FRIEND
Clinical otolaryngology : official journal of ENT-UK ; official journal of Netherlands Society for Oto-Rhino-Laryngology & Cervico-Facial Surgery
Testing for laryngopharyngeal reflux in exhaled breath condensate: a pilot study.
PMID 23253346