EMAIL THIS PAGE TO A FRIEND
Current protocols in nucleic acid chemistry
Rapid and efficient synthesis of nucleoside polyphosphates and their conjugates using sulfonyl imidazolium salts.
PMID 23255202
Abstract
This unit describes a method for preparing nucleoside polyphosphates and their conjugates such as nucleoside triphosphates, symmetrical and unsymmetrical dinucleoside polyphosphates and sugar nucleotides. The protocols employ sulfonyl imidazolium salts (SnISs) as coupling reagents. These reagents are prepared in two steps in very high yield from commercially available materials and can be stored for at least 1 year at -20°C. The tetra-n-butylammonium salts of nucleoside mono-, di-, or triphosphates are reacted with the SnISs to form reactive phosphoryl imidazolium donors. These donors are reacted with the tetra-n-butylammonium salts of pyrophosphate, nucleotide mono- or diphosphates or sugar-1-phosphates to give the nucleoside polyphosphates and their conjugates in excellent yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
270547
N,N-Dimethylformamide, for HPLC, ≥99.9%
C3H7NO
319937
N,N-Dimethylformamide, ACS reagent, ≥99.8%
C3H7NO
33120
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
C3H7NO
D158550
N,N-Dimethylformamide, ReagentPlus®, ≥99%
C3H7NO
494488
N,N-Dimethylformamide, biotech. grade, ≥99.9%
C3H7NO
651664
Tetrabutylammonium azide
C16H36N4
86868
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
C16H37NO4S
86853
Tetrabutylammonium bisulfate, for ion pair chromatography, LiChropur, ≥99.0%
C16H37NO4S
98479
Tetrabutylammonium bisulfate solution, for ion pair chromatography, LiChropur, concentrate, ampule
C16H37NO4S
72453
Tetrabutylammonium bisulfate solution, ~55% in H2O
C16H37NO4S
426288
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
C16H36BrN
193119
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
C16H36BrN
86857
Tetrabutylammonium bromide, for ion pair chromatography, LiChropur, ≥99.0%
C16H36BrN
462144
Tetrabutylammonium bromide solution, 50 wt. % in H2O
C16H36BrN
86870
Tetrabutylammonium chloride, ≥97.0% (NT)
C16H36ClN
86852
Tetrabutylammonium chloride, for ion pair chromatography, LiChropur, ≥99.0%
C16H36ClN
358665
Tetrabutylammonium cyanide, 95%
C17H36N2
79989
Tetrabutylammonium cyanide, technical, ≥80%
C17H36N2
216143
Tetrabutylammonium fluoride solution, 1.0 M in THF
C16H36FN
361399
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
C16H36FN
155837
Tetrabutylammonium hydrogensulfate, 97%
C16H37NO4S
86854
Tetrabutylammonium hydroxide solution, ~40% in water, for ion chromatography
C16H37NO
230189
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
C16H37NO
178780
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
C16H37NO
86880
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
C16H37NO
86863
Tetrabutylammonium hydroxide solution, 54.0-56.0% in H2O
C16H37NO
140775
Tetrabutylammonium iodide, reagent grade, 98%
C16H36IN
86890
Tetrabutylammonium iodide, ≥99.0% (AT)
C16H36IN
86903
Tetrabutylammonium iodide, for ion pair chromatography, LiChropur, ≥99.0%
C16H36IN
359173
Tetrabutylammonium nitrate, 97%
C16H36N2O3