EMAIL THIS PAGE TO A FRIEND
Journal of experimental botany
Towards establishing broad-spectrum disease resistance in plants: silicon leads the way.
PMID 23255278
Abstract
Plants are constantly threatened by a wide array of microbial pathogens. Pathogen invasion can lead to vast yield losses and the demand for sustainable plant-protection strategies has never been greater. Chemical plant activators and selected strains of rhizobacteria can increase resistance against specific types of pathogens but these treatments are often ineffective or even cause susceptibility against others. Silicon application is one of the scarce examples of a treatment that effectively induces broad-spectrum disease resistance. The prophylactic effect of silicon is considered to be the result of both passive and active defences. Although the phenomenon has been known for decades, very little is known about the molecular basis of silicon-afforded disease control. By combining knowledge on how silicon interacts with cell metabolism in diatoms and plants, this review describes silicon-induced regulatory mechanisms that might account for broad-spectrum plant disease resistance. Priming of plant immune responses, alterations in phytohormone homeostasis, regulation of iron homeostasis, silicon-driven photorespiration and interaction with defence signalling components all are potential mechanisms involved in regulating silicon-triggered resistance responses. Further elucidating how silicon exerts its beneficial properties may create new avenues for developing plants that are better able to withstand multiple attackers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
343250
Silicon, pieces, 99.95% trace metals basis
Si
267414
Silicon, powder, −60 mesh, 99.999% trace metals basis
Si
215619
Silicon, powder, −325 mesh, 99% trace metals basis
Si
633097
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Si
646687
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Si
647535
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Si
647780
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Si
647802
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm
Si
647675
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Si
647764
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Si
647101
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Si
647543
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Si
647799
Silicon, wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm
Si
647705
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Si
647772
Silicon, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Si
767492
Silicon, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.999% trace metals basis
Si
GF37993595
Silicon, rod, 100mm, diameter 10.0mm, single crystal - random orientation, 100%
Si
GF64686998
Silicon, rod, 100mm, diameter 2.0mm, crystalline, 100%
Si
GF22929307
Silicon, rod, 100mm, diameter 5.0mm, crystalline, 100%
Si
GF16363391
Silicon, rod, 10mm, diameter 2.0mm, crystalline, 100%
Si
GF66465276
Silicon, rod, 25mm, diameter 2.0mm, crystalline, 100%
Si
GF43555664
Silicon, rod, 25mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF70438518
Silicon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Si
GF35274113
Silicon, rod, 40mm, diameter 20mm, single crystal, -111, 99.999%
Si
GF05482852
Silicon, rod, 50mm, diameter 2.0mm, crystalline, 100%
Si
GF91515034
Silicon, rod, 50mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF14076930
Silicon, rod, 50mm, diameter 5.0mm, crystalline, 100%
Si
GF28448691
Silicon, rod, 50mm, diameter 5mm, single crystal, -100, 99.999%
Si
GF40945754
Silicon, rod, 50mm, diameter 5mm, single crystal, -111, 99.999%
Si
GF64342188
Silicon, rod, 50mm, diameter 6mm, single crystal, -111, 99.999%
Si