EMAIL THIS PAGE TO A FRIEND
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Double and quadruple tetracycline labeling of bone: impact of the label itself.
PMID 23255455