EMAIL THIS PAGE TO A FRIEND
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova
[The comparative study on the efficacy of the combination of serotonin reuptake inhibitor antidepressants and antipsychotics in the treatment of recurrent depressive disorders].
PMID 23257758
Abstract
A combination of serotonin reuptake inhibitor antidepressants (prozac and stimulaton) with atypical antipsychotics (zyprexa and solian) reduced depression in patients with recurrent depressive disorders during 10 days. The effect was evenly distributed between 10, 20 and 40 days of treatment. Other symptoms had a peculiar dynamics depending on the therapy. By the end of the study, similar effects were achieved for all groups. The addition of antipsychotics to antidepressant treatment insignificantly increased the number of adverse events.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8010
(±)-Sulpiride
C15H23N3O4S