EMAIL THIS PAGE TO A FRIEND
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition
Bioactive anti-obesity food components.
PMID 23258394
Abstract
Overweight and obesity are the epidemics of the 21st century in developed countries. Obesity is associated with several metabolic disorders and various diseases such as type 2 diabetes, cardiovascular disease, high blood pressure, dyslipidemia (high levels of circulating triacylglycerols and low-density lipoprotein cholesterol, and low levels of high-density lipoprotein cholesterol), commonly referred to as Metabolic Syndrome (MS). Although there is a therapeutic arsenal to combat some of these diseases, especially type 2 diabetes and cardiovascular disease, both the intake of proper diets and maintaining healthy lifestyles are considered the best preventive measures. The general population has access to a variety of dietary foods for weight loss, and to nutritional supplements. It is important to identify those foods and supplements that are effective and safe. The purpose of this review is to examine, from a critical point of view, food components currently used by health professionals and by the general population as coadjuvants in the prevention of obesity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2378
γ-Linolenic acid, ≥99%, liquid
C18H30O2
62174
γ-Linolenic acid, analytical standard
C18H30O2