EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Elucidating combinatorial histone modifications and crosstalks by coupling histone-modifying enzyme with biotin ligase activity.
PMID 23258705
Abstract
Histone post-translational modifications (PTMs) often form complex patterns of combinations and cooperate to specify downstream biological processes. In order to systemically analyse combinatorial PTMs and crosstalks among histone PTMs, we have developed a novel nucleosome purification method called Biotinylation-assisted Isolation of CO-modified Nucleosomes (BICON). This technique is based on physical coupling of the enzymatic activity of a histone-modifying enzyme with in vivo biotinylation by the biotin ligase BirA, and using streptavidin to purify the co-modified nucleosomes. Analysing the nucleosomes isolated by BICON allows the identification of PTM combinations that are enriched on the modified nucleosomes and function together within the nucleosome context. We used this new approach to study MSK1-mediated H3 phosphorylation and found that MSK1 not only directly phosphorylated H3, but also induced hyperacetylation of both histone H3 and H4 within the nucleosome. Moreover, we identified a novel crosstalk pathway between H3 phosphorylation and H4 acetylation on K12. Involvement of these acetyl marks in MSK1-mediated transcription was further confirmed by chromatin immunoprecipitation assays, thus validating the biological relevance of the BICON results. These studies serve as proof-of-principle for this new technical approach, and demonstrate that BICON can be further adapted to study PTMs and crosstalks associated with other histone-modifying enzymes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-911
Anti-acetyl-Histone H3 (Lys14) Antibody, Use Anti-acetyl-Histone H3 (Lys14) Antibody (Rabbit Polyclonal Antibody) validated in ICC, IP, WB to detect acetyl-Histone H3 (Lys14) also known as H3K14Ac, Histone H3 (acetyl K14).
06-599
Anti-acetyl-Histone H3 Antibody, Detect acetyl-Histone H3 with Anti-acetyl-Histone H3 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in WB, ICC, ChIP, ChIP-seq.
07-595
Anti-acetyl-Histone H4 (Lys12) Antibody, Use Anti-acetyl-Histone H4 (Lys12) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, DB, WB, ChIP-seq to detect acetyl-Histone H4 (Lys12) also known as H4K12Ac, Histone H4 (acetyl K12).
07-329
Anti-acetyl-Histone H4 (Lys16) Antibody, Anti-acetyl-Histone H4 (Lys16) Antibody is a rabbit polyclonal antibody for detection of acetyl-Histone H4 (Lys16) also known as H4K16Ac, Histone H4 (acetyl K16 & has been published and validated in ChIP, WB, Mplex, PIA, DB, ChIP-seq.
05-858
Anti-Histone H4 Antibody, pan, clone 62-141-13, rabbit monoclonal, Anti-Histone H4 Antibody, pan, clone 62-141-13 is a Rabbit Monoclonal Antibody for detection of Histone H4 also known as Histone H4 & has been validated in ChIP, WB, Mplex, ChIP-seq.
06-946
Anti-hyperacetylated Histone H4 (Penta) Antibody, Use Anti-hyperacetylated Histone H4 (Penta) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, ICC, WB to detect hyperacetylated Histone H4 (Penta) also known as H4 histone family member A.
H9908
Monoclonal Anti-phospho-Histone H3 (pSer28) antibody produced in rat, ~0.5 mg/mL, clone HTA28, purified immunoglobulin, buffered aqueous solution
12-370
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.