EMAIL THIS PAGE TO A FRIEND
Journal of Translational Medicine
Blockade of phospholipid scramblase 1 with its N-terminal domain antibody reduces tumorigenesis of colorectal carcinomas in vitro and in vivo.
PMID 23259795