EMAIL THIS PAGE TO A FRIEND
International journal of clinical pharmacology and therapeutics
Multi-targeted polycyclic polyprenylated acylphloroglucinols are major constituents of Cuban propolis and contributors to its anticancer activity.
PMID 23260000