EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Novel in vivo active anti-malarials based on a hydroxy-ethyl-amine scaffold.
PMID 23260352
Abstract
A novel series of anti-malarials, based on a hydroxy-ethyl-amine scaffold, initially identified as peptidomimetic protease inhibitors is described. Combination of the hydroxy-ethyl-amine anti-malarial phramacophore with the known Mannich base pharmacophore of amodiaquine (57) resulted in promising in vivo active novel derivatives.