EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Experimental and theoretical study of molecular structure of beryllium, magnesium, calcium, strontium and barium 4-nitrobenzoates.
PMID 23261035
Abstract
The influence of alkaline earth metal ions on the electronic system of 4-nitrobenzoic acid was studied in this paper. The vibrational (FT-IR) and NMR ((1)H and (13)C) spectra were recorded for 4-nitrobenzoic acid (4-nba) and its salts (4-nb). The assignment of vibrational spectra was done. Some shifts of band wavenumbers in alkaline earth metal 4-nitrobenzoates spectra were observed in the series from magnesium to barium salts. Good correlations between wavenumbers of the vibrational bands in the IR spectra of studied salts and ionic potential, electronegativity, inverse of atomic mass, ionic radius and ionization energy of studied metals were found. The regular changes in the chemical shifts of protons ((1)H NMR) and carbons ((13)C NMR) in the series of studied salts were also observed. Optimized geometrical structures of studied compounds were calculated by B3LYP method using 6-311++G(**) as well as LANL2DZ basis sets. Theoretical wavenumbers and intensities in IR and chemical shifts in NMR spectra were also obtained. The calculated parameters were compared with experimental data of studied compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72910
4-Nitrobenzoic acid, purum, ≥98.0% (HPLC)
C7H5NO4
461091
4-Nitrobenzoic acid, 98%
C7H5NO4