EMAIL THIS PAGE TO A FRIEND
Pakistan journal of pharmaceutical sciences
Carbopol 934, 940 and Ultrez 10 as viscosity modifiers of palm olein esters based nano-scaled emulsion containing ibuprofen.
PMID 23261730
Abstract
Micro-emulsions and sometimes nano-emulsions are well known candidates to deliver drugs locally. However, the poor rheological properties are marginally affecting their acceptance pharmaceutically. This work aimed to modify the poor flow properties of a nano-scaled emulsion comprising palm olein esters as the oil phase and ibuprofen as the active ingredient for topical delivery. Three Carbopol ® resins: 934, 940 and Ultrez 10, were utilized in various concentrations to achieve these goals. Moreover, phosphate buffer and triethanolamine solutions pH 7.4 were used as neutralizing agents to assess their effects on the gel-forming and swelling properties of Carbopol ® 940. The addition of these polymers caused the produced nano-scaled emulsion to show a dramatic droplets enlargement of the dispersed globules, increased intrinsic viscosity, consistent zeta potential and transparent-to-opaque change in appearance. These changes were relatively influenced by the type and the concentration of the resin used. Carbopol ® 940 and triethanolamine appeared to be superior in achieving the proposed tasks compared to other materials. The higher the pH of triethanolamine solution, the stronger the flow-modifying properties of Carbopol ® 940. Transmission electron microscopy confirmed the formation of a well-arranged gel network of Carbopol ® 940, which was the major cause for all realized changes. Later in vitro permeation studies showed a significant decrease in the drug penetration, thus further modification using 10% w/w menthol or limonene as permeation promoters was performed. This resulted in in vitro and in vivo pharmacodynamics properties that are comparably higher than the reference chosen for this study.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63670
(±)-Menthol, racemic, ≥98.0% (GC)
C10H20O
62118
(R)-(+)-Limonene, analytical standard
C10H16
62120
(R)-(+)-Limonene, purum, ≥96.0% (sum of enantiomers, GC)
C10H16
62122
(R)-(+)-Limonene, technical, ~90% (sum of enantiomers, GC)
C10H16
W266507
DL-Menthol, ≥95%, FCC, FG
C10H20O
M2772
Menthol, 99%
C10H20O
81117
Poly(acrylic acid sodium salt), analytical standard, for GPC, 1,250
81123
Poly(acrylic acid sodium salt), analytical standard, for GPC, 16,000
81131
Poly(acrylic acid sodium salt), analytical standard, for GPC, 83,000
420344
Poly(acrylic acid sodium salt), average Mw ~2,100
447013
Poly(acrylic acid sodium salt), average Mw ~5,100 by GPC, powder
181293
Poly(acrylic acid), analytical standard, average Mn 130,000 (Typical)
323667
Poly(acrylic acid)
181285
Poly(acrylic acid), average Mv ~450,000
306215
Poly(acrylic acid), average Mv ~1,250,000
306223
Poly(acrylic acid), average Mv ~3,000,000
306231
Poly(acrylic acid), average Mv ~4,000,000
535931
Poly(acrylic acid) solution, average Mw ~2,000, 50 wt. % in H2O, electronic grade
523925
Poly(acrylic acid) solution, average Mw ~100,000, 35 wt. % in H2O
416002
Poly(acrylic acid) solution, average Mw ~250,000, 35 wt. % in H2O
416010
Poly(acrylic acid, sodium salt) solution, average Mw ~1,200, 45 wt. % in H2O
416029
Poly(acrylic acid, sodium salt) solution, average Mw ~8,000, 45 wt. % in H2O
416037
Poly(acrylic acid, sodium salt) solution, average Mw ~15,000, 35 wt. % in H2O
T1502
Triethanolamine hydrochloride, ≥99.5% (titration)
C6H15NO3 · HCl
T9534
Triethanolamine hydrochloride, BioXtra, ≥99.5% (titration)
C6H15NO3 · HCl