EMAIL THIS PAGE TO A FRIEND
Regional anesthesia and pain medicine
Propylene glycol and glycerol concentration in ultrasound gel.
PMID 23263219
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
540242
(R)-(−)-1,2-Propanediol, 96%
C3H8O2
540250
(S)-(+)-1,2-Propanediol, 96%
C3H8O2