EMAIL THIS PAGE TO A FRIEND
Ecotoxicology (London, England)
The response of soil organism communities to the application of the insecticide lindane in terrestrial model ecosystems.
PMID 23264019
Abstract
The EU plant protection regulation 1107/2009/EC defines the requirements for active ingredients to be approved, specifically including the assessment of effects on biodiversity and ecosystems. According to that, semi-field methods are expected to be more important in the near future. Therefore, a higher-tier experiment suitable to assess the risk for soil organisms was conducted to further develop the TME (terrestrial model ecosystems) methodology in a dose-response design with the persistent insecticidal model compound lindane (gamma-HCH). The effects of lindane on soil communities such as collembolans, oribatid mites, nematodes, soil fungi and plant biomass were determined in 42 TME. Intact TME-soil cores (diameter 300 mm, height 400 mm) from undisturbed grassland were stored outdoor under natural climatic conditions. Lindane was applied in five concentrations between 0.032 mg active ingredients (ai)/kg dry soil and 3.2 mg ai/kg dry weight soil, six-fold replicated each. Twelve TME served as untreated controls. Abundance and community structures of oribatids, collembolans, enchytraeids, nematodes and fungi were recorded. Oribatid mites' community responded 3 months after treatment, although they were not significantly affected by the overall treatment regimen. Collembolans in total and species-specific abundance as well as the community endpoints (principal response curves, diversity measures) were adversely affected by moderate dosages of lindane. Effects were transient between 3 and 5 months after treatment with a recovery within 1 year. No significant effects could be detected for enchytraeids, nematodes and fungi. The study design and the obtained results allow for calculations of no observed effect concentrations below the highest treatment level for populations and for soil communities as defined entities, as well as effective concentrations. The paper discusses the limits of effect detection in the light of achievable coefficients of variation and by means of minimum detectable differences. Outdoor TME are useful to analyze and assess functional and structural endpoints in soil organisms' communities and their possible recovery after pesticide treatment within 1 year.