EMAIL THIS PAGE TO A FRIEND
Journal of dental research
Effect of local vs. systemic bisphosphonate delivery on dental implant fixation in a model of osteonecrosis of the jaw.
PMID 23264610
Abstract
Locally applied bisphosphonates may improve the fixation of metal implants in bone. However, systemic bisphosphonate treatment is associated with a risk of osteonecrosis of the jaw (ONJ). We hypothesized that local delivery of bisphosphonate from the implant surface improves the fixation of dental implants without complications in a setting where systemic treatment induces ONJ. Forty rats were randomly allocated to 4 groups of 10. All groups received a titanium implant inserted in an extraction socket. Group I received the implants only. Group II received dexamethasone (0.5 mg/kg). Group III received dexamethasone as above plus alendronate (200 µg/kg). Group IV received zoledronate-coated implants and dexamethasone as above. The animals were sacrificed 2 weeks after tooth extraction. All 10 animals with systemic alendronate treatment developed large ONJ-like changes, while all with local treatment were completely healed. Implant removal torque was higher for the bisphosphonate-coated implants compared with the other groups (p < 0.03 for each comparison). Micro-computed tomography of the maxilla showed more bone loss in the systemic alendronate group compared with groups receiving local treatment (p = 0.001). Local bisphosphonate treatment appears to improve implant fixation in a setting where systemic treatment caused ONJ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4978
Alendronate sodium trihydrate, ≥97% (NMR), powder
C4H12NaNO7P2 · 3 H2O