EMAIL THIS PAGE TO A FRIEND
Pediatric dentistry
Heart rate, salivary α-amylase activity, and cooperative behavior in previously naïve children receiving dental local anesthesia.
PMID 23265159
Abstract
Psychosomatic indicators, such as heart rate (HR), salivary alpha amylase (sAA) activity, and behavior, can be used to determine stress. This study's aim was to assess the pattern of changes of salivary alpha amylase, heart rate, and cooperative behavior in previously naïve children receiving dental treatment under local anesthesia. Included were 30 children with no prior dental experience who needed 4 or more sessions of dental treatment involving local anesthesia. In each session, sAA, HR, and behavior were assessed before and during the application of local anesthesia and at the end of the treatment. The highest sAA value was always observed at the end of each session; overall, the value was lower in the fourth session. HR always increased during the local anesthesia, and did not vary across sessions. No significant relationship was found between child cooperation and either sAA or HR. In this sample, child cooperation may not be an accurate indicator of stress. Based on salivary alpha amylase activity changes, dental treatment involving local anesthesia in naïve children appeared to be less stressful after 3 sessions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8220
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
10065 α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
86250 α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
A9857
α-Amylase from Aspergillus oryzae, powder, ≥150 units/mg protein (biuret)
A7595
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
A3403
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
A4582 α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
A4551
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
A4862 α-Amylase from Bacillus licheniformis, liquid
A6814 α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
A6380 α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
10069 α-Amylase from Bacillus subtilis, powder, yellow-brown, ~380 U/mg
10070 α-Amylase from Bacillus subtilis, powder, yellow-brown, ~50 U/mg
A9972
α-Amylase from human pancreas, lyophilized powder, ≥100 units/mg protein
A1031
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
A0521
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
A6255
α-Amylase from porcine pancreas, DFP Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
A4268
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
A3306
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
86247 Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg