EMAIL THIS PAGE TO A FRIEND
Burns : journal of the International Society for Burn Injuries
Is there still a need for silver sulfadiazine in small burns?
PMID 23265279
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
481181
Silver(I) sulfadiazine, 98%
C10H9AgN4O2S