EMAIL THIS PAGE TO A FRIEND
Medical hypotheses
Linking respiratory chain uncoupling to amyotrophic lateral sclerosis implies potential treatment with herbal extracts containing genipin.
PMID 23265356
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G4796
Genipin, ≥98% (HPLC), powder
C11H14O5