EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Methodology for the validation of analytical methods involved in uniformity of dosage units tests.
PMID 23265732
Abstract
Validation of analytical methods is required prior to their routine use. In addition, the current implementation of the Quality by Design (QbD) framework in the pharmaceutical industries aims at improving the quality of the end products starting from its early design stage. However, no regulatory guideline or none of the published methodologies to assess method validation propose decision methodologies that effectively take into account the final purpose of developed analytical methods. In this work a solution is proposed for the specific case of validating analytical methods involved in the assessment of the content uniformity or uniformity of dosage units of a batch of pharmaceutical drug products as proposed in the European or US pharmacopoeias. This methodology uses statistical tolerance intervals as decision tools. Moreover it adequately defines the Analytical Target Profile of analytical methods in order to obtain analytical methods that allow to make correct decisions about Content uniformity or uniformity of dosage units with high probability. The applicability of the proposed methodology is further illustrated using an HPLC-UV assay as well as a near infra-red spectrophotometric method.