EMAIL THIS PAGE TO A FRIEND
Journal of photochemistry and photobiology. B, Biology
Study on the synthesis and properties of mussel mimetic poly(ethylene glycol) bioadhesive.
PMID 23266052
Abstract
The catecholic amino acid 3,4-Dihydroxyphenyl-l-alanine (DOPA) is believed to be responsible for the water-resistant adhesive characteristics of mussel adhesive proteins (MAPs). In this paper, the synthesis of three-armed poly(ethylene glycol)s with dopamine endgroups (PEG-DOPAs) using acylation reaction and Michael addition reaction was described, and PEG-DOPAs was used as bioadhesive under the UV irradiation. The structures of PEG-DOPAs were characterized by FTIR, (1)H NMR spectroscopy. The adhesion properties of PEG-DOPAs were investigated and results were significantly superior to 0.05MPa of the commercially available fibrin adhesives. Moreover, the adhesive strength of PEG-DOPAs (Mw 3000) increased from 0.587MPa to 1.82MPa, PEG-DOPAs (Mw 20,000) increased from 0.338MPa to 1.92MPa with the elongation of the binding time, severally. The photopolymerization kinetic study was taken to evaluate gelation time of bioadhesive indirectly. The cytotoxicity of photocured PEG-DOPAs hydrogels for L929 cells was evaluated by the fluorescence microscopy and MTT (3-[4-dimethylthiazol-2-yl]-2,5-diphenyltetrazolium bromide; Thiazolyl blue) assay, respectively. The results showed that PEG-DOPAs were biocompatible and less cytotoxicity toward the growth of L929 cells. The swelling properties of PEG-DOPAs hydrogels were also tested. PEG-DOPAs (3000) reached swelling equilibrium within 2h at 37°C, and PEG-DOPAs (20,000) within 6h at 37°C.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9503
3,4-Dihydroxy-DL-phenylalanine
C9H11NO4