EMAIL THIS PAGE TO A FRIEND
Chemosphere
Oxidative stress and arsenic toxicity: role of NADPH oxidases.
PMID 23266413
Abstract
The effect of arsenic (25 and 50 μM As for 1 and 5d) was analysed in wild type (WT) and Arabidopsis thaliana (L.) Heynh plants deficient in NADPH oxidase C (AtrbohC). The content of H(2)O(2) and malondialdehyde (MDA) increased with the As concentration, while the opposite effect was found for NO in WT and AtrbohC plants. The As treatment reduced catalase and increased glutathione reductase activities to the same extent in WT and AtrbohC plants, although the induction of all SOD isoforms (mainly CuZn-SODs) was observed in WT plants, the opposite effects being found in AtrbohC plants. Glycolate oxidase (H(2)O(2) producers) considerably increased with the concentration and time of treatment with As in WT and AtrbohC mutants. Arsenic induced the uptake and translocation of P, S, Cu, Zn, and Fe in WT plants, while in AtrbohC plants the opposite trend was noted and the uptake of As became considerably lower than in WT plants. These results suggest that As causes oxidative stress by inducing glycolate oxidase, while NADPH oxidase does not appear to participate in ROS overproduction but could be critical in regulating antioxidant defences as well as the transport and translocation of As and macro/micronutrients.