EMAIL THIS PAGE TO A FRIEND
Ultrasonics sonochemistry
Removal of Brilliant Green from wastewater using conventional and ultrasonically prepared poly(acrylic acid) hydrogel loaded with kaolin clay: A comparative study.
PMID 23266437
Abstract
The present work deals with the removal of Brilliant Green dye from wastewater using a poly(acrylic acid) hydrogel composite (PAA-K hydrogel) prepared by incorporation of kaoline clay. The composite has been synthesized using ultrasound assisted polymerization process as well as the conventional process, with an objective of showing the better effectiveness of ultrasound assisted synthesis. It has been observed that the hydrogel prepared by ultrasound assisted polymerization process showed better results. The optimum conditions for the removal of dye are pH of 7, temperature of 35°C, initial dye concentration of 30mg/L and hydrogel loading of 1g. The extent of removal of dye increased with an increase in the contact time and initial dye concentration. A pseudo-second-order kinetic model has been developed to explain the adsorption kinetics of dye on the PAA-K hydrogel. Thermodynamic and kinetic parameters indicate that the adsorption process is spontaneous in nature and the PAA-K hydrogel prepared by ultrasound process is a promising adsorbent compared to conventional process. The obtained adsorption data has also been fitted into commonly used adsorption isotherms and it has been found that Freundlich as well as Langmuir adsorption isotherm models fits well to the experimental results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B6756
Brilliant Green, Dye content ~90 %
C27H34N2O4S
B4014
Brilliant Green, certified by the Biological Stain Commission
C27H34N2O4S
685445
Halloysite nanoclay
H4Al2O9Si2 · 2 H2O
K7375 Kaolin
18616 Kaolin, heavy, powder
K1512 Kaolin, meets USP testing specifications
60609 Kaolin, tested according to Ph.Eur.
17-0010 Kaolin, CP
81117
Poly(acrylic acid sodium salt), analytical standard, for GPC, 1,250
81123
Poly(acrylic acid sodium salt), analytical standard, for GPC, 16,000
81131
Poly(acrylic acid sodium salt), analytical standard, for GPC, 83,000
420344
Poly(acrylic acid sodium salt), average Mw ~2,100
447013
Poly(acrylic acid sodium salt), average Mw ~5,100 by GPC, powder
181293
Poly(acrylic acid), analytical standard, average Mn 130,000 (Typical)
323667
Poly(acrylic acid)
181285
Poly(acrylic acid), average Mv ~450,000
306215
Poly(acrylic acid), average Mv ~1,250,000
306223
Poly(acrylic acid), average Mv ~3,000,000
306231
Poly(acrylic acid), average Mv ~4,000,000
535931
Poly(acrylic acid) solution, average Mw ~2,000, 50 wt. % in H2O, electronic grade
523925
Poly(acrylic acid) solution, average Mw ~100,000, 35 wt. % in H2O
416002
Poly(acrylic acid) solution, average Mw ~250,000, 35 wt. % in H2O
416010
Poly(acrylic acid, sodium salt) solution, average Mw ~1,200, 45 wt. % in H2O
416029
Poly(acrylic acid, sodium salt) solution, average Mw ~8,000, 45 wt. % in H2O
416037
Poly(acrylic acid, sodium salt) solution, average Mw ~15,000, 35 wt. % in H2O