EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Nickel-catalyzed cross couplings of benzylic ammonium salts and boronic acids: stereospecific formation of diarylethanes via C-N bond activation.
PMID 23268734
Abstract
We have developed a nickel-catalyzed cross coupling of benzylic ammonium triflates with aryl boronic acids to afford diarylmethanes and diarylethanes. This reaction proceeds under mild reaction conditions and with exceptional functional group tolerance. Further, it transforms branched benzylic ammonium salts to diarylethanes with excellent chirality transfer, offering a new strategy for the synthesis of highly enantioenriched diarylethanes from readily available chiral benzylic amines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
539775
Ethane, 99.99%
C2H6
295302
Ethane, ≥99%
C2H6